Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLF13 Rabbit anti-Human, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501732131 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-2131 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-2131 Supplier Proteintech Group Inc Supplier No. 183521AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-RANTES factor of Late Activated T Lymphocytes-1 (RFLAT-1) polyclonal antiserum. RFLAT-1 is a novel protein belonging to the TFIIIA-like zinc finger protein superfamily and binds to the A site of the RANTES (Regulated upon Activation, Normal T cell Expressed and Secreted) chemokine. RANTES is involved in the generation of inflammatory infiltrate and HIV entry into immune cells.

Specifications

Antigen KLF13
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene KLF13
Gene Accession No. Q9Y2Y9
Gene Alias BTE binding protein 3, BTEB3, FKLF2, KLF13, Krueppel like factor 13, Kruppel like factor 13, NSLP1, RFLAT 1, RFLAT1, Transcription factor BTEB3, Transcription factor NSLP1
Gene Symbols KLF13
Host Species Rabbit
Immunogen KLF13 Fusion Protein Ag13200
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 499171, 51621
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.