Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLF14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100880
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100880 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100880 Supplier Invitrogen™ Supplier No. PA5100880
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total KLF14.

This gene encodes a member of the Kruppel-like family of transcription factors. The gene exhibits imprinted expression from the maternal allele in embryonic and extra-embryonic tissues. Expression of this gene is induced by TGF-beta, and the protein represses TGF-beta receptor II expression. The protein functions as part of a transcriptional co-repressor complex that also contains the transcriptional regulator SIN3A and histone deacetylase 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLF14
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene KLF14
Gene Accession No. Q19A41, Q8TD94
Gene Alias 5330411L03Rik; AL022736; Basic transcription element-binding protein 5; BTEB5; BTE-binding protein 5; KLF14; Krueppel-like factor 14; Kruppel like factor 14; Kruppel-like factor 14; transcription factor BTEB5
Gene Symbols KLF14
Host Species Rabbit
Immunogen A synthesized peptide derived from human KLF14(Accession Q8TD94), corresponding to amino acid residues C260-G310.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 136259, 619665
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.