Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLF14 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP311925100UG
Description
KLF14 Polyclonal antibody specifically detects KLF14 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
KLF14 | |
Polyclonal | |
Western Blot 1 - 2 ug/ml, Immunohistochemistry 2 - 4 ug/ml | |
basic transcription element-binding protein 5, Kruppel-like factor 14 | |
Synthetic peptide derived from the human KLF14 protein (Uniprot: Q8TD94) | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
Unconjugated | |
PBS | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
136259 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction