Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLF15 Monoclonal Antibody (2G8)
GREENER_CHOICE

Catalog No. PIMA515491
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515491 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515491 Supplier Invitrogen™ Supplier No. MA515491
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15491 targets KLF15 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15491 immunogen is purified recombinant fragment of human KLF15 expressed in E. Coli. MA5-15491 detects KLF15 which has a predicted molecular weight of approximately 44kDa.

KLF15 is a nuclear protein belonging to the Sp1 C2H2-type zinc-finger protein family with three C2H2-type zinc fingers. A transcriptional activator, KLF15 binds to the GA element of the CLCNKA promoter and may help to regulate the strict tissue and nephron segment-specific expression of CLCNKA and CLCNKB channel genes. It plays an essential role in adipogenesis in cells through its regulation of PPARG expression. Reports suggest that KLF15 is an inhibitor of cardiac hypertrophy and functions in a pathway, regulating the cardiac response to hemodynamic stress, and regulates mitochondrial acetyl-CoA synthase expression in skeletal muscle. KLF15 may also act as a transcriptional repressor of the rhodopsin and IRBP promoters in the retina, and is a possible participant in repression of photoreceptor-specific gene expression in nonphotoreceptor cells. It is highly expressed in liver, followed by heart, skeletal muscle, and kidney but not expressed in bone marrow or lymphoid tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLF15
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2G8
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.03% sodium azide
Gene KLF15
Gene Accession No. Q9UIH9
Gene Alias 1810013I09Rik; AV048136; AW494632; Cardiovascular Krueppel-like factor; CKLF; DKFZp779M1320; Kidney-enriched krueppel-like factor; kidney-enriched Kruppel-like factor; KKLF; KKLF protein; Klf15; KLF15; transcription factor; Krueppel-like factor 15; Kruppel like factor 15; Kruppel-like factor 15
Gene Symbols KLF15
Host Species Mouse
Immunogen Purified recombinant fragment of human KLF15 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28999
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.