Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLF16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518057
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518057 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518057 Supplier Invitrogen™ Supplier No. PA518057
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

Transcription factor that binds GC and GT boxes and displaces Sp1 and Sp3 from these sequences. Modulates dopaminergic transmission in the brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLF16
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene KLF16
Gene Accession No. Q9BXK1
Gene Alias AI843742; basic transcription element binding protein 4; basic transcription element-binding protein 4; BTEB4; BTE-binding protein 4; dopamine receptor regulating factor; dopamine receptor-regulating factor; Drrf; Klf16; Krueppel-like factor 16; Kruppel like factor 16; Kruppel-like factor 16; novel Sp1-like zinc finger transcription factor 2; NSLP2; transcription factor BTEB4; Transcription factor NSLP2
Gene Symbols KLF16
Host Species Goat
Immunogen Synthetic peptide sequence (PAPSPAPSPAPAGL) corresponding to the C-terminus amino acids of KLF16 (aa 239-252).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83855
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.