Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLF16 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | KLF16 |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
KLF16 Polyclonal specifically detects KLF16 in Human samples. It is validated for Western Blot.Specifications
KLF16 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
basic transcription element binding protein 4, Basic transcription element-binding protein 4, BTEB4Transcription factor NSLP2, dopamine receptor regulating factor, DRRF, Krueppel-like factor 16, Kruppel-like factor 16, Novel Sp1-like zinc finger transcription factor 2, NSLP2BTE-binding protein 4, Transcription factor BTEB4 | |
The immunogen is a synthetic peptide directed towards the middle region of human KLF16 (NP_114124). Peptide sequence LAKHARRHPGFHPDLLRRPGARSTSPSDSLPCSLAGSPAPSPAPSPAPAG | |
Affinity purified |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS buffer, 2% sucrose | |
83855 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title