Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLF17 Antibody (PCRP-KLF17-1G2), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314000PE
This item is not returnable.
View return policy
Description
KLF17 Monoclonal antibody specifically detects KLF17 in Human samples. It is validated for Flow CytometrySpecifications
| KLF17 | |
| Monoclonal | |
| PE | |
| FLJ40160, Kruppel-like factor 17, Zfp393, Zinc finger protein 393Krueppel-like factor 17, ZNF393novel zinc-finger protein | |
| Recombinant full-length human KLF17protein (Uniprot: Q5JT82) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| PCRP-KLF17-1G2 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 128209 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction