Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLF17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589403
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589403 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589403 Supplier Invitrogen™ Supplier No. PA589403
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QMLPPQDAHD LGMPPAESQS LLVLGSQDSL VSQPDSQEGP FLPEQPGPAP QTVEKNSRPQ EGTGRRGSSE ARPYCCNYEN CGKAYTKRSH LVSHQRKHTG ERPYSCNWES CSWSFFRSDE LRRHMRVHTR YRPYKCDQCS REFMRSDHLK QHQKTHRPGP SDPQANNNNG EQDSPPAAGP; Positive Samples: U-87MG, HepG2, MCF7, Mouse brain; Cellular Location: Nucleus.

KLF17 is a transcription repressor that binds to the promoter of target genes and prevents their expression. Acts as a negative regulator of epithelial-mesenchymal transition and metastasis in breast cancer. Specifically binds the 5'-CACCC-3' sequence in the promoter of ID1, a key metastasis regulator in breast cancer, and repress its expression. May be a germ cell-specific transcription factor that plays important roles in spermatid differentiation and oocyte development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLF17
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.53 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene KLF17
Gene Accession No. Q5JT82, Q8CFA7
Gene Alias 7420700M05Rik; AA420409; AU043468; C85123; D4Ertd561e; FLJ40160; germ cell specific zinc finger protein; germ cell-specific zinc finger protein; Gzf; KLF17; Krueppel-like factor 17; Kruppel like factor 17; Kruppel-like factor 17; novel zinc-finger protein; Zfp393; Zinc finger protein 393; ZLF393; ZNF393
Gene Symbols KLF17
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 210-389 of human KLF17 (NP_7757553).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 128209, 75753
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.