Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLF2 Antibody (665333), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB5466F
Description
KLF2 Monoclonal antibody specifically detects KLF2 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOFSpecifications
KLF2 | |
Monoclonal | |
FITC | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
10365 | |
Store at 4°C in the dark. | |
IgG2b |
Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
665333 | |
Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
Krueppel-like factor 2, Kruppel-like factor 2 (lung), Kruppel-like factor LKLF, LKLFLung krueppel-like factor, lung Kruppel-like zinc finger transcription factor | |
E. coli-derived recombinant human KLF2, Pro71-Pro168 (predicted), Accession # Q9Y5W3 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction