Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLF2 Antibody (OTI3A10), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271597B
Description
KLF2 Monoclonal specifically detects KLF2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
KLF2 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
KLF2 | |
Full length recombinant protein of human KLF2 (NP_057354) produced in E.coli. | |
0.1 mL | |
10365.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI3A10 | |
Western Blot, Immunohistochemistry | |
Krueppel-like factor 2, Kruppel-like factor 2 (lung), Kruppel-like factor LKLF, LKLFLung krueppel-like factor, lung Kruppel-like zinc finger transcription factor | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction