Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLF2 Antibody (OTI3A10), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP245510
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry
KLF2 Monoclonal specifically detects KLF2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
KLF2 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000, Immunohistochemistry 1:150, Immunohistochemistry-Paraffin | |
Krueppel-like factor 2, Kruppel-like factor 2 (lung), Kruppel-like factor LKLF, LKLFLung krueppel-like factor, lung Kruppel-like zinc finger transcription factor | |
Mouse | |
37.2 kDa | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
OTI3A10 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
KLF2 | |
Full length human recombinant protein of human KLF2 (NP_057354) produced in E.coli. | |
Immunogen affinity purified | |
RUO | |
10365 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction