Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLF2 Monoclonal Antibody (2F12A11)
GREENER_CHOICE

Catalog No. FERMA548691
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548691 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548691 Supplier Invitrogen™ Supplier No. MA548691
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KLF2 Monoclonal Antibody for Western Blot, ELISA

KLF2 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 3 C2H2-type zinc fingers. KLF2 binds to the CACCC box in the beta-globin gene promoter and activates transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLF2
Applications ELISA, Western Blot, Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 2F12A11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene KLF2
Gene Accession No. Q60843, Q9Y5W3
Gene Alias KLF2; Krueppel-like factor 2; Kruppel like factor 2; Kruppel-like factor 2; Kruppel-like factor 2 (lung); Kruppel-like factor LKLF; Lklf; LOW QUALITY PROTEIN: Krueppel-like factor 2; lung krueppel-like factor; lung Kruppel-like factor; lung Kruppel-like zinc finger transcription factor
Gene Symbols KLF2
Host Species Mouse
Immunogen Purified recombinant fragment of human KLF2 (AA: 251-355) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10365, 16598
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.