Learn More
KLF2 Mouse anti-Human, Clone: OTI3D12, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF806992

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
KLF2 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 3 C2H2-type zinc fingers. KLF2 binds to the CACCC box in the beta-globin gene promoter and activates transcription.Specifications
KLF2 | |
Monoclonal | |
Unconjugated | |
KLF2 | |
LKLF | |
Mouse | |
Affinity Chromatography | |
RUO | |
10365 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI3D12 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9Y5W3 | |
KLF2 | |
Full length human recombit protein of human KLF2 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.