Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLF2 Polyclonal Antibody, Cyanine5, Bioss
SDP

Catalog No. 502279864
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-9864 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-9864 Supplier Bioss Supplier No. BS2772RCY5
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KLF2 Polyclonal antibody specifically detects KLF2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)

Specifications

Antigen KLF2
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Cyanine5
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene KLF2
Gene Accession No. Q60843, Q9ET58, Q9Y5W3
Gene Alias KLF2; Krueppel-like factor 2; Kruppel like factor 2; Kruppel-like factor 2; Kruppel-like factor 2 (lung); Kruppel-like factor LKLF; Lklf; LOW QUALITY PROTEIN: Krueppel-like factor 2; lung krueppel-like factor; lung Kruppel-like factor; lung Kruppel-like zinc finger transcription factor
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human KLF2
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10365, 16598, 306330
Target Species Human, Mouse, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.