Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLF3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530621
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530621 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530621 Supplier Invitrogen™ Supplier No. PA530621
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A549, HCT116. Predicted reactivity: Human (99%), Mouse (98%), Rat (99%), Pig (99%), Rhesus Monkey (99%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Krueppel-like factors (KLFs) comprise a family of evolutionarily conserved zinc finger-containing transcription factors with diverse regulatory functions in cell growth, proliferation, differentiation and embryogenesis. Individual members of the Sp1-like/KLF family can function either as activators or repressors, depending on which promoter they bind and the coregulators with which they interact. KLF6, also designated Zf9 or CPBP (core promoter-binding protein), and KLF3 are Kruppel-like zinc finger containing transcription factors. KLF6 is rapidly induced during hepatic stellate cell activation and transactivates a reporter gene driven by the collagen I promoter, suggesting KLF6 plays a role in the response to tissue injury. KLF3 may play a role in hematopoiesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLF3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene Klf3
Gene Accession No. P57682
Gene Alias 9930027G08Rik; AL024007; Basic krueppel-like factor; basic kruppel like factor; Basic Kruppel- like factor; basic Kruppel-like factor; BKLF; CACCC-box-binding protein BKLF; klf12; Klf3; Krueppel-like factor 3; kruppel like factor 12; Kruppel like factor 3; Kruppel-like factor 12; Kruppel-like factor 3 (basic); MGC48279; TEF-2; transcript ch138; zinc finger containing transcription factor
Gene Symbols Klf3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human KLF3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51274
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.