Learn More
Invitrogen™ KLF3 Polyclonal Antibody

Description
Immunogen sequence: LMFDPVPVKQE AMDPVSVSYP SNYMESMKPN KYGVIYSTPL PEKFFQTPEG LSHGIQMEPV DL Highest antigen sequence identity to the following orthologs - mouse 92%, rat 94%.
Specifications
Specifications
| Antigen | KLF3 |
| Applications | ChIP Assay, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Klf3 |
| Gene Accession No. | P57682 |
| Gene Alias | 9930027G08Rik; AL024007; Basic krueppel-like factor; basic kruppel like factor; Basic Kruppel- like factor; basic Kruppel-like factor; BKLF; CACCC-box-binding protein BKLF; klf12; Klf3; Krueppel-like factor 3; kruppel like factor 12; Kruppel like factor 3; Kruppel-like factor 12; Kruppel-like factor 3 (basic); MGC48279; TEF-2; transcript ch138; zinc finger containing transcription factor |
| Gene Symbols | Klf3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.