Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLF4 Antibody (3A1), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | KLF4 |
---|---|
Clone | 3A1 |
Dilution | Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:20-1:200 |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry |
Classification | Monoclonal |
Description
KLF4 Monoclonal antibody specifically detects KLF4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, ImmunocytochemistrySpecifications
KLF4 | |
Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Epithelial zinc finger protein EZF, EZFendothelial Kruppel-like zinc finger protein, GKLFKrueppel-like factor 4, Gut-enriched krueppel-like factor, Kruppel-like factor 4 (gut) | |
A synthesized peptide derived from Human KLF4 [UniProt O43474] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
3A1 | |
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
9314 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title