Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLF4 Antibody (4E6) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP121047 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP121047 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP121047 Supplier Novus Biologicals Supplier No. NBP121047
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KLF4 Monoclonal specifically detects KLF4 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.

Specifications

Antigen KLF4
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 4000000
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Flow Cytometry 1x10^6cells, ELISA 1:100-1:2000
Gene Accession No. O43474
Gene Alias Epithelial zinc finger protein EZF, EZFendothelial Kruppel-like zinc finger protein, GKLFKrueppel-like factor 4, Gut-enriched krueppel-like factor, Kruppel-like factor 4 (gut)
Gene Symbols KLF4
Host Species Mouse
Immunogen Recombinant human KLF4 (1-170aa) purified from E. coli (NP_004226.3).
Molecular Weight of Antigen 55 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9314
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.