Learn More
Invitrogen™ KLF4 Polyclonal Antibody

Description
This antibody is predicted to react with bovine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
Specifications
Specifications
| Antigen | KLF4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | KLF4 |
| Gene Accession No. | O43474 |
| Gene Alias | endothelial Kruppel-like zinc finger protein; epithelial zinc finger protein EZF; Ezf; Gklf; gut Kruppel-like factor; gut-enriched krueppel-like factor; high mobility group nucleosomal binding domain 3; high mobility group nucleosome-binding domain-containing protein 3; HMGN3; KLF4; Krueppel-like factor 4; Kruppel like factor 4; Kruppel-like factor 4 (Gut enriched kruppel-like factor) (Epithelial zinc-finger protein EZF); Kruppel-like factor 4 (gut); PNAS-24; PNAS-25; RP11-150J11.1; thyroid hormone receptor interacting protein 7; Thyroid receptor-interacting protein 7; TR-interacting protein 7; TRIP7; Zie |
| Gene Symbols | KLF4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.