Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KLF4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA523184
Description
KLF4 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Kruppel like factor 4 (KLF4) protein is encoded by the KLF4 gene located on chromosome 9 in humans. It is also known as gut-enriched Kruppel-like factor (GKLF) or epithelial zinc-finger protein (EZF) and belongs to a family of conserved Kruppel-like factor family of DNA binding zinc finger transcription factors. KLF4 exerts its functional relevance across varied cellular pathways. As one of the four Yamanaka factors, it plays a key role in reprogramming differentiated cells into pluripotent stem cells. Most studies have focused on its specific role in endothelial cell signaling, adipogenesis, differentiation, embryogenesis and cell cycle control. Localization plays an important role in the biological significance of this protein wherein nuclear localization is associated with its function as an oncogene and cytosolic localization with tumor suppression specific effect.
Specifications
KLF4 | |
Polyclonal | |
Unconjugated | |
KLF4 | |
endothelial Kruppel-like zinc finger protein; epithelial zinc finger protein EZF; Ezf; Gklf; gut Kruppel-like factor; gut-enriched krueppel-like factor; high mobility group nucleosomal binding domain 3; high mobility group nucleosome-binding domain-containing protein 3; HMGN3; KLF4; Krueppel-like factor 4; Kruppel like factor 4; Kruppel-like factor 4 (Gut enriched kruppel-like factor) (Epithelial zinc-finger protein EZF); Kruppel-like factor 4 (gut); PNAS-24; PNAS-25; RP11-150J11.1; thyroid hormone receptor interacting protein 7; Thyroid receptor-interacting protein 7; TR-interacting protein 7; TRIP7; Zie | |
Rabbit | |
Protein G | |
RUO | |
114505, 16600, 9314 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
O43474, Q60793 | |
KLF4 | |
A portion of amino acids 1-50 of human KLF4 was used as the immunogen for this antibody. | |
100 μg | |
Primary | |
Human, Mouse, Rat, Monkey | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction