Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLF5 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298939C
Description
KLF5 Polyclonal specifically detects KLF5 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
KLF5 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
Basic transcription element-binding protein 2, BTEB2, BTE-binding protein 2, CKLFGC-box-binding protein 2, Colon krueppel-like factor, colon kruppel-like factor, GC box binding protein 2, IKLFbasic transcription element binding protein 2, Intestinal-enriched krueppel-like factor, intestinal-enriched kruppel-like factor, Krueppel-like factor 5, Kruppel-like factor 5 (intestinal), Transcription factor BTEB2 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human KLF5. | |
0.1 mL | |
Cancer | |
688 | |
Store at 4°C in the dark. | |
IgG |
Immunofluorescence | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction