Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLF5 Monoclonal Antibody (OTI1E4), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA811878

Description
KLF5 is a member of the Kruppel-like factor subfamily of zinc finger proteins. It contains three C2H2-type zinc fingers at the carboxyl-terminus that preferentially bind to cis-DNA elements that are GC rich. KLF5 is a transcription activator that promotes cell proliferation and oncogenesis.Specifications
KLF5 | |
Monoclonal | |
Unconjugated | |
KLF5 | |
BTEB2, CKLF, IKLF | |
Mouse | |
Affinity Chromatography | |
RUO | |
688 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
OTI1E4 | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q13887 | |
KLF5 | |
Human recombit protein fragment corresponding to amino acids 1-457 of human KLF5 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction