Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLF5 Rabbit anti-Human, Mouse, Unconjugated, Polyclonal, Active Motif™
SDP

Catalog No. 501993141
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3141 100 μL
50-199-3142 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3141 Supplier Active Motif Supplier No. 61099
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KLF5 is a member of the Kruppel-like factor subfamily of zinc finger proteins. It contains three C2H2-type zinc fingers at the carboxyl-terminus that preferentially bind to cis-DNA elements that are GC rich. KLF5 is a transcription activator that promotes cell proliferation and oncogenesis.

Specifications

Antigen KLF5
Applications ChIP Assay, ChIP sequencing (ChIP-seq), Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
Gene KLF5
Gene Accession No. Q13887, Q9Z0Z7
Gene Alias (intestinal Kruppel-like factor; 4930520J07Rik; basic transcription element binding protein 2; basic transcription element binding protein BTEB2; Basic transcription element-binding protein 2; Bteb2; BTE-binding protein 2; CKLF; Colon krueppel-like factor; colon kruppel-like factor; GC box binding protein 2; GC-box-binding protein 2; Iklf; Intestinal-enriched krueppel-like factor; intestinal-enriched kruppel-like factor; Klf5; Klf5C isoform; Krueppel-like factor 5; Kruppel like factor 5; Kruppel-like factor 5; Kruppel-like factor 5 (intestinal); RP11-505F3.5; Transcription factor BTEB2
Gene Symbols KLF5
Host Species Rabbit
Immunogen This kLF5 antibody was raised against a peptide derived from the N-terminus of mouse kLF5 (krueppel-like factor 5).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12224, 688
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.