Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLF5 Recombinant Rabbit Monoclonal Antibody (HL2269)
GREENER_CHOICE

Catalog No. FEMA550468
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550468 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550468 Supplier Invitrogen™ Supplier No. MA550468
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

KLF5 Recombinant Monoclonal Antibody for Western Blot, IHC (P)

KLF5 is a member of the Kruppel-like factor subfamily of zinc finger proteins. It contains three C2H2-type zinc fingers at the carboxyl-terminus that preferentially bind to cis-DNA elements that are GC rich. KLF5 is a transcription activator that promotes cell proliferation and oncogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLF5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone HL2269
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene KLF5
Gene Accession No. Q13887, Q9Z0Z7
Gene Alias (intestinal Kruppel-like factor; 4930520J07Rik; basic transcription element binding protein 2; basic transcription element binding protein BTEB2; basic transcription element-binding protein 2; BTEB2; BTE-binding protein 2; CKLF; colon krueppel-like factor; colon kruppel-like factor; GC box binding protein 2; GC-box-binding protein 2; IKLF; Intestinal-enriched krueppel-like factor; intestinal-enriched kruppel-like factor; Klf5; Klf5C isoform; Krueppel-like factor 5; Kruppel like factor 5; Kruppel-like factor 5; Kruppel-like factor 5 (intestinal); RP11-505F3.5; transcription factor BTEB2
Gene Symbols KLF5
Host Species Rabbit
Immunogen Recombinant fragment of human KLF5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12224, 688, 84410
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.