Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLF6 Antibody [CoraFluor™ 1], R&D Systems™
SDP

Catalog No. AF3499CL1 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3499CL1 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AF3499CL1 Supplier R&D Systems Supplier No. AF3499CL1

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Sheep Polyclonal Antibody

KLF6 Polyclonal antibody specifically detects KLF6 in Human samples. It is validated for Western Blot

Specifications

Antigen KLF6
Applications Western Blot
Classification Polyclonal
Conjugate CoraFluor 1
Formulation PBS
Gene Alias BCD1Zf9, B-cell-derived protein 1, COPEBCBA1, core promoter element binding protein, Core promoter element-binding protein, CPBPZF9, DKFZp686N0199, EC 2.1.1.43, EC 6.1.1.15, GBF, GC-rich binding factor, GC-rich sites-binding factor GBF, Krueppel-like factor 6, Kruppel-like factor 6, Kruppel-like zinc finger protein Zf9, PAC1, Proto-oncogene BCD1, protooncogene B-cell derived 1, ST12, suppression of tumorigenicity 12 (prostate), Suppressor of tumorigenicity 12 protein, Transcription factor Zf9
Host Species Sheep
Immunogen E. coli-derived recombinant human KLF6, Met1-Ser109, Accession # Q99612
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Prostate Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1316
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.