Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLF6 Monoclonal Antibody (9A2)
GREENER_CHOICE

Catalog No. 396900
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-690-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 39-690-0 Supplier Invitrogen™ Supplier No. 396900
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

KLF6 Monoclonal Antibody for Western Blot, ICC/IF, IHC, ELISA

This gene encodes a member of the Kruppel-like family of transcription factors. The zinc finger protein is a transcriptional activator, and functions as a tumor suppressor. Multiple transcript variants encoding different isoforms have been found for this gene, some of which are implicated in carcinogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLF6
Applications ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 9A2
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene Klf6
Gene Accession No. Q99612
Gene Alias Aa1017; AI448727; BCD1; B-cell-derived protein 1; C86813; CBA1; Copeb; core promoter element binding protein; core promoter element-binding protein; CPBP; FM2; FM6; GBF; GC-rich binding factor; GC-rich sites-binding factor GBF; Ierepo1; Ierepo3; immediate early response, erythropoietin 1; KLF6; Krueppel-like factor 6; Kruppel like factor 6; Kruppel-like factor 6; Kruppel-like zinc finger protein Zf9; PAC1; proto-oncogene BCD1; protooncogene B-cell derived 1; R75280; ST12; suppression of tumorigenicity 12 (prostate); suppressor of tumorigenicity 12 protein; Transcription factor Zf9; ZF9
Gene Symbols Klf6
Host Species Mouse
Immunogen Recombinant protein derived from the internal region of the human KLF6 splice variant 1.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1316
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.