Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLF6 Mouse anti-Human, Clone:4C7D6, Abnova™

Catalog No. 89333526 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-526 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-526 Supplier Abnova Corporation Supplier No. MAB17290
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human KLF6.

This gene encodes a member of the Kruppel-like family of transcription factors. The zinc finger protein is a transcriptional activator, and functions as a tumor suppressor. Multiple transcript variants encoding different isoforms have been found for this gene, some of which are implicated in carcinogenesis. [provided by RefSeq]

Specifications

Antigen KLF6
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4C7D6
Conjugate Unconjugated
Description Kruppel-like factor 6
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene KLF6
Gene Alias BCD1, COPEB, CPBP, DKFZp686N0199, GBF, PAC1, ST12, ZF9
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 71-283 of of human KLF6 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 1316
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.