Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLF6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579560
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579560 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579560 Supplier Invitrogen™ Supplier No. PA579560
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Human Placenta Tissue, Rat Testis Tissue, HELA whole cell, HEPG2 whole cell, HEPA whole cell.

This gene encodes a member of the Kruppel-like family of transcription factors. The zinc finger protein is a transcriptional activator, and functions as a tumor suppressor. Multiple transcript variants encoding different isoforms have been found for this gene, some of which are implicated in carcinogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLF6
Applications Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide
Gene Klf6
Gene Accession No. O35819, Q99612
Gene Alias Aa1017; AI448727; BCD1; B-cell-derived protein 1; C86813; CBA1; Copeb; core promoter element binding protein; core promoter element-binding protein; CPBP; FM2; FM6; GBF; GC-rich binding factor; GC-rich sites-binding factor GBF; Ierepo1; Ierepo3; immediate early response, erythropoietin 1; KLF6; Krueppel-like factor 6; Kruppel like factor 6; Kruppel-like factor 6; Kruppel-like zinc finger protein Zf9; PAC1; proto-oncogene BCD1; protooncogene B-cell derived 1; R75280; ST12; suppression of tumorigenicity 12 (prostate); suppressor of tumorigenicity 12 protein; Transcription factor Zf9; ZF9
Gene Symbols Klf6
Host Species Rabbit
Immunogen E.coli-derived human KLF6 recombinant protein (Position: E38-N205).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1316, 58954
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.