Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLF6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5104348

Description
Antibody detects endogenous levels of total KLF6.
This gene encodes a member of the Kruppel-like family of transcription factors. The zinc finger protein is a transcriptional activator, and functions as a tumor suppressor. Multiple transcript variants encoding different isoforms have been found for this gene, some of which are implicated in carcinogenesis.
Specifications
KLF6 | |
Polyclonal | |
Unconjugated | |
Klf6 | |
Aa1017; AI448727; BCD1; B-cell-derived protein 1; C86813; CBA1; Copeb; core promoter element binding protein; core promoter element-binding protein; CPBP; FM2; FM6; GBF; GC-rich binding factor; GC-rich sites-binding factor GBF; Ierepo1; Ierepo3; immediate early response, erythropoietin 1; KLF6; Krueppel-like factor 6; Kruppel like factor 6; Kruppel-like factor 6; Kruppel-like zinc finger protein Zf9; PAC1; proto-oncogene BCD1; protooncogene B-cell derived 1; R75280; ST12; suppression of tumorigenicity 12 (prostate); suppressor of tumorigenicity 12 protein; Transcription factor Zf9; ZF9 | |
Rabbit | |
Affinity chromatography | |
RUO | |
1316, 23849, 58954 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O08584, O35819, Q99612 | |
Klf6 | |
A synthesized peptide derived from human KLF6(Accession Q99612), corresponding to amino acid residues P149-V199. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction