Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLF6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501729667 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-9667 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-9667 Supplier Proteintech Group Inc Supplier No. 147161AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody raise against the full length of KLF6 gene of human origin, and can recognize both of KLF6, included KLF6 (∽32 kDa), p-KLF6(∽35-46 kDa). Otherwise, there may be a non-specific band(50 kDa) shown.

This gene encodes a member of the Kruppel-like family of transcription factors. The zinc finger protein is a transcriptional activator, and functions as a tumor suppressor. Multiple transcript variants encoding different isoforms have been found for this gene, some of which are implicated in carcinogenesis.

Specifications

Antigen KLF6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene KLF6
Gene Accession No. O08584, O35819, Q99612
Gene Alias B cell derived protein 1, COPEB, CPBP, GBF, KLF6, Krueppel like factor 6, Kruppel like factor 6, PAC1, Proto oncogene BCD1, ST12, Transcription factor Zf9, ZF9
Gene Symbols Klf6
Host Species Rabbit
Immunogen KLF6 Fusion Protein Ag6440
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1316, 23849, 58954
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.