Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLF7 Antibody (3G3), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242626IR
Description
KLF7 Monoclonal specifically detects KLF7 in Human samples. It is validated for Western Blot, ELISA.Specifications
KLF7 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
KLF7 | |
Recombinant human KLF7 (1-302aa) purified from E. coli | |
100 μL | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
3G3 | |
Western Blot, ELISA | |
Krueppel-like factor 7, Kruppel-like factor 7 (ubiquitous), ubiquitous Kruppel-like transcription factor, UKLFUbiquitous krueppel-like factor | |
Mouse | |
Protein A purified | |
RUO | |
8609 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction