Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLF7 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299217APC
Description
KLF7 Polyclonal antibody specifically detects KLF7 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin)Specifications
KLF7 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
Krueppel-like factor 7, Kruppel-like factor 7 (ubiquitous), ubiquitous Kruppel-like transcription factor, UKLFUbiquitous krueppel-like factor | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human KLF7. | |
0.1 mL | |
Neuroscience | |
8609 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction