Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLF7 Monoclonal Antibody (OTI3F6), OriGene

Mouse Monoclonal Antibody
Supplier: OriGene CF812003


Description
KLF7 Monoclonal antibody specifically detects KLF7 in Human samples. It is validated for Western BlotSpecifications
KLF7 | |
Monoclonal | |
See Label | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q2KHT4 | |
Mouse | |
Protein A/G | |
RUO | |
83445 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Western Blot | |
OTI3F6 | |
Unconjugated | |
GSG1 | |
9830124P08Rik; Klf7; Krueppel-like factor 7; Kruppel like factor 7; Kruppel-like factor 7 (ubiquitous); Ubiquitous krueppel-like factor; ubiquitous Kruppel-like factor; ubiquitous Kruppel-like transcription factor; UKLF | |
Human recombinant protein fragment corresponding to amino acids 43-246 of human GSG1 (NP_112579) produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction