Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLF7 Monoclonal Antibody (OTI8D7), OriGene
SDP

Catalog No. 502543846
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3846 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3846 Supplier OriGene Supplier No. CF812009
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KLF7 Monoclonal antibody specifically detects KLF7 in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen KLF7
Applications Western Blot
Classification Monoclonal
Clone OTI8D7
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene Klf7
Gene Accession No. O75840, Q99JB0
Gene Alias 9830124P08Rik; Klf7; Krueppel-like factor 7; Kruppel like factor 7; Kruppel-like factor 7 (ubiquitous); Ubiquitous krueppel-like factor; ubiquitous Kruppel-like factor; ubiquitous Kruppel-like transcription factor; UKLF
Host Species Mouse
Immunogen Full length human recombinant protein of human KLF7 (NP_003700) produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8609, 93691
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.