Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLF7 Mouse anti-Human, Mouse, Clone: OTI8D7, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA812009

Description
The protein encoded by this gene is a member of the Kruppel-like transcriptional regulator family. Members in this family regulate cell proliferation, differentiation and survival and contain three C2H2 zinc fingers at the C-terminus that mediate binding to GC-rich sites. This protein may contribute to the progression of type 2 diabetes by inhibiting insulin expression and secretion in pancreatic beta-cells and by deregulating adipocytokine secretion in adipocytes. A pseudogene of this gene is located on the long arm of chromosome 3. Alternative splicing results in multiple transcript variants.Specifications
KLF7 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
O75840, Q99JB0 | |
KLF7 | |
Full length human recombit protein of human KLF7 produced in E.coli. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Western Blot | |
OTI8D7 | |
Unconjugated | |
KLF7 | |
UKLF | |
Mouse | |
Affinity Chromatography | |
RUO | |
8609, 93691 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction