Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLF7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109884
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109884 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109884 Supplier Invitrogen™ Supplier No. PA5109884
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SETFGEDLDC FLHASPPPCI EESFRRLDPL LLPVEAAICE KSSAVDILLS RDKLLSETCL SLQPASSSLD SYTAVNQAQL NAVTSLTPPS SPELSRHLVK TSQTLSAVDG TVTLKLVAKK AALSSVKVRS LISAHGRDVS GVLHEAMSSR GTTGNTQVQS PSNATTATGV FPGLTILPST.

The protein encoded by this gene is a member of the Kruppel-like transcriptional regulator family. Members in this family regulate cell proliferation, differentiation and survival and contain three C2H2 zinc fingers at the C-terminus that mediate binding to GC-rich sites. This protein may contribute to the progression of type 2 diabetes by inhibiting insulin expression and secretion in pancreatic beta-cells and by deregulating adipocytokine secretion in adipocytes. A pseudogene of this gene is located on the long arm of chromosome 3. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLF7
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene Klf7
Gene Accession No. O75840, Q99JB0
Gene Alias 9830124P08Rik; Klf7; Krueppel-like factor 7; Kruppel like factor 7; Kruppel-like factor 7 (ubiquitous); Ubiquitous krueppel-like factor; ubiquitous Kruppel-like factor; ubiquitous Kruppel-like transcription factor; UKLF
Gene Symbols Klf7
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 51-230 of human KLF7 (NP_001257871.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8609, 93691
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.