Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLF8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01674645
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-645 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-645 Supplier Invitrogen™ Supplier No. PA19061
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with porcine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

This gene encodes a protein which is a member of the Sp/KLF family of transcription factors. Members of this family contain a C-terminal DNA-binding domain with three Kruppel-like zinc fingers. The encoded protein is thought to play an important role in the regulation of epithelial to mesenchymal transition, a process which occurs normally during development but also during metastasis. A pseudogene has been identified on chromosome 16. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLF8
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene KLF8
Gene Accession No. O95600
Gene Alias A830097P10Rik; Basic krueppel-like factor 3; basic kruppel-like factor 3; BKLF3; DKFZp686O08126; DXS741; KLF8; Klf8-ps1; Krueppel-like factor 8; Kruppel like factor 8; Kruppel-like factor 8; Kruppel-like factor 8, pseudogene 1; LOW QUALITY PROTEIN: Krueppel-like factor 8; MGC138314; RGD1565831; RP13-1021K9.1; zinc finger protein 741; ZNF74; ZNF741
Gene Symbols KLF8
Host Species Goat
Immunogen Synthetic peptide corresponding to the N terminus amino acids VDMDKLINNLEVQ (aa 2-15).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11279
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.