Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLF9 Antibody (OTI8A11) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270285
Description
KLF9 Monoclonal specifically detects KLF9 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
KLF9 | |
Monoclonal | |
LYOPH | |
Western Blot 1:2000, Immunohistochemistry 1:150 | |
basic transcription element binding protein 1, Basic transcription element-binding protein 1, BTEB1GC-box-binding protein 1, BTEBBTE-binding protein 1, Krueppel-like factor 9, Kruppel-like factor 9, Transcription factor BTEB1 | |
Mouse | |
27.1 kDa | |
100 μg | |
687 | |
Human, Mouse, Rat | |
Lyophilized |
Western Blot, Immunohistochemistry | |
OTI8A11 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
KLF9 | |
Full length human recombinant protein of human KLF9(NP_001197) produced in E.coli. | |
Affinity Purified | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction