Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLF9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117090
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117090 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117090 Supplier Invitrogen™ Supplier No. PA5117090
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: A-549, K-562, HepG2, Mouse brain, Mouse kidney, Rat brain Immunogen sequence: MSAAAYMDFV AAQCLVSISN RAAVPEHGVA PDAERLRLPE REVTKEHGDP GDTWKDYCTL VTIAKSLLDL NKYRPIQTPS VCSDSLESPD EDMGSDSDVT TESGSSPSHS PEERQDPGSA PSPLSLLHPG VAAKGKHASE KRHKCPYSGC.

KLF-9 is a transcription factor belonging to the Sp 1 C2H2 type zinc finger family. It binds to GC box elements located in the promoter. This binding of the encoded protein to a single GC box inhibits mRNA expression while binding to tandemly repeated GC box elements activates transcription. KLF-9 serves as a key transcriptional regulator for uterine endometrial cell proliferation, adhesion, and differentiation both during pregnancy and tumorigenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLF9
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.86 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene KLF9
Gene Accession No. O35739, Q01713, Q13886
Gene Alias 2310051E17Rik; AA589643; basic transcription element binding protein 1; basic transcription element-binding protein 1; BTEB; BTEB1; BTEB-1; BTE-binding protein 1; ENSMUSG00000055440; GC-box-binding protein 1; Gm9971; KLF9; Krueppel-like factor 9; Kruppel like factor 9; Kruppel-like factor 9; transcription factor BTEB1
Gene Symbols KLF9
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human KLF9 (NP_0011971).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117560, 16601, 687
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.