Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLH Antibody (3B1), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP244049
Description
Ensure accurate, reproducible results in Western Blot, ELISA, Immunohistochemistry
KLH Monoclonal specifically detects KLH in All Species samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
KLH | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000-1:10000, ELISA 1:20000-1:100000, Immunohistochemistry 1:1000-1:5000 | |
keyhole limpet hemocyanin (KLH) | |
KLH Antibody (3B1) was produced in mouse by repeated immunizations with KLH (Keyhole Limpet Hemocyanin) followed by hybridoma development. | |
0.1 mg | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgM |
Western Blot, ELISA, Immunohistochemistry | |
3B1 | |
Unconjugated | |
0.1 M Tris Chloride, 0.5 M Sodium Chloride, pH 8.0 with 0.1% Sodium Azide | |
Mouse | |
Immunogen affinity purified | |
RUO | |
All species | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction