Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLH Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299936AF350
Description
KLH Polyclonal specifically detects KLH in Non-species specific samples. It is validated for ELISA.Specifications
KLH | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Non-species specific | |
Purified |
ELISA | |
Alexa Fluor 350 | |
keyhole limpet hemocyanin (KLH) | |
Produced in rabbits immunized with purified, recombinant Keyhole Limpet Hemocyanin. | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction