Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLH Antibody, Alexa Fluor™ 532 , Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP278810AF532
Description
KLH Polyclonal antibody specifically detects KLH in Nonspecies specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, ImmunofluorescenceSpecifications
KLH | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Non-species specific | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence | |
Alexa Fluor 532 | |
keyhole limpet hemocyanin (KLH) | |
KLH was used as the immunogen. | |
1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction