Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLH Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1207054
Description
KLH Polyclonal specifically detects KLH in All Species samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
KLH | |
Polyclonal | |
Unconjugated | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
Rabbit | |
Affinity Purified | |
RUO | |
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Rabbit Serum. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1 mg/ml | |
Western Blot 1:1000-1:5000, ELISA 1:10000-1:50000, Immunohistochemistry 1:500-1:2500, Immunohistochemistry-Paraffin 1:10-1:500, Fluorophore-linked immunosorbent assay | |
keyhole limpet hemocyanin (KLH) | |
KLH (Keyhole Limpet Hemocyanin) | |
0.2 mg | |
Primary | |
All species | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction