Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLH Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299936PCP
Description
KLH Polyclonal antibody specifically detects KLH in Nonspecies specific samples. It is validated for ELISASpecifications
KLH | |
Polyclonal | |
ELISA | |
keyhole limpet hemocyanin (KLH) | |
Produced in rabbits immunized with purified, recombinant KLH (Keyhole Limpet Hemocyanin). | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG |
ELISA | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Non-species specific | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction