Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLH Monoclonal Antibody (3B1), Rockland™
SDP

Catalog No. 501990599
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0599 25 μL
50-199-0598 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-0599 Supplier Rockland Immunochemicals Supplier No. 200301E66S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. KLH antibody is specific for KLH. Does not cross react with mouse heart or skin tissue sections. Cross-reactivity from other sources has not been determined.

Specifications

Antigen KLH
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 3B1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris HCl with 0.5M NaCl and 0.1% sodium azide; pH 8
Host Species Mouse
Immunogen Anti-KLH (MOUSE) Monoclonal Antibody was produced in mouse by repeated immunizations with KLH (Keyhole Limpet Hemocyanin) followed by hybridoma development.
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgM
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.