Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLHDC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554215
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554215 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554215 Supplier Invitrogen™ Supplier No. PA554215
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FLCGGLSADN IPLSDGWIHN VTTNCWKQLT HLPKTRPRLW HTACLGKENE IMVFGGSKDD LLALDTGHCN DLLIFQTQPY SLLRSCLDCI GKNSIMLESQ ISLLPPKLLQ QVLKKITFWA AANHREEQRV Q Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 88%.

KLHDC1 contains 6 Kelch repeats and is widely expressed, with high levels in skeletal muscle, pancreas and liver. KLHDC1 and KLHDC2 have differential localization and activity in cultured mammalian cells. The exact function of KLHDC1 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLHDC1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KLHDC1
Gene Accession No. Q8N7A1
Gene Alias kelch domain containing 1; kelch domain-containing protein 1; Klhdc1; MST025; MSTP025
Gene Symbols KLHDC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KLHDC1. Recombinant protein control fragment (Product #RP-88913).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 122773
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.