Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLHDC10 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA554155

Catalog No. PIPA554155


Only null left
Add to Cart

Description

Description

Immunogen sequence: EWTQLKPNNL SCDLPEERYR HEIAHDGQRI YILGGGTSWT AYSLNKIHAY NLETNAWEEI ATKPHEKIGF PAARRCHSCV QIK Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

KLHDC10
Polyclonal
Unconjugated
KLHDC10
kelch domain containing 10; kelch domain-containing protein 10; KIAA0265; KLHDC10; PNAS-119; RGD1309792; scruin like at the midline homolog; slim
Rabbit
Antigen Affinity Chromatography
RUO
23008
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q6PID8
KLHDC10
Recombinant protein corresponding to Human KLHDC10. Recombinant protein control fragment (Product #RP-92715).
100 μL
Primary
Human, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.