Learn More
Invitrogen™ KLHDC2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551428
Description
Immunogen sequence: MADGNEDLRA DDLPGPAFES YESMELACPA ERSGHVAVSD GRHMFVWGGY KSNQVRGLYD FYLPREELWI YNMETGRWKK INTEGDVPPS MSGSCAVCVD RVLYLFGGHH SRGNTNKFYM LDSRSTDRVL QWERIDCQGI PPSSKDKL Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
Represses CREB3-mediated transcription by interfering with CREB3-DNA binding. [UniProt].
Specifications
KLHDC2 | |
Polyclonal | |
Unconjugated | |
Klhdc2 | |
2310022K15Rik; D12Ertd522e; endothelial differentiation inhibitory protein TNG; HCA33; HCLP1; HCLP-1; Hepatocellular carcinoma-associated antigen 33; host cell factor homolog LCP; Host cell factor-like protein 1; kelch domain containing 2; kelch domain-containing protein 2; KLHDC2; LCP; similar to kelch domain containing 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23588 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunohistochemistry | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y2U9 | |
Klhdc2 | |
Recombinant protein corresponding to Human KLHDC2. Recombinant protein control fragment (Product #RP-88886). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.