Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLHDC5 Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318445FR
Description
KLHDC5 Polyclonal antibody specifically detects KLHDC5 in Human, Mouse samples. It is validated for Western BlotSpecifications
KLHDC5 | |
Polyclonal | |
Western Blot | |
Ctb9, kelch domain containing 5, kelch domain-containing protein 5, KIAA1340, MGC131714 | |
Recognizes region between residues Between Between 400 and 450.. | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
57542 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction