Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KLHDC7B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116314
Description
KLHDC7B Polyclonal Antibody for Western Blot
Specifications
| KLHDC7B | |
| Polyclonal | |
| Unconjugated | |
| KLHDC7B | |
| EG546648; Gm306; kelch domain containing 7B; kelch domain-containing protein 7B; KLHDC7B; RGD1562257 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 113730 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.46 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q96G42 | |
| KLHDC7B | |
| KLH conjugated synthetic peptide between 442-471 amino acids from the C-terminal region of human KLHDC7B. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction